Merck
  • A somaclonal variant of rose-scented geranium (Pelargonium spp.) with moderately high content of isomenthone in its essential oil.
A somaclonal variant of rose-scented geranium (Pelargonium spp.) with moderately high content of isomenthone in its essential oil.
Natural product communications (2012-10-19)
Swaroop S Kulkarni, Nagawara S Ravindra, Kalavagunta V N S Srinivas, Raghavendra N Kulkarni
ZUSAMMENFASSUNG
Rose-scented geranium (Pelargonium spp.), which is highly valued for its essential oil, is exclusively propagated vegetatively. Hence no genetic improvement work is possible through conventional breeding. Somaclonal variation was generated with and without in vitro mutagenesis using N-nitroso-N-methyl urea (NMU) in an Indian cultivar 'Bourbon', and a clone 'Narmada'. A somaclonal variant (N75) with a moderately high content of isomenthone in its essential oil was isolated from somaclones generated after treatment of internodal explants of clone, 'Narmada' with 0.25 mM NMU for 1 h. The contents of isomenthone in its essential oil were 26% and 35%, respectively, in SC2/VM2 and SC3/VM3 generations (second and third vegetative generations, respectively, after in vitro mutagen treatment) as compared with 0.7% and 0.3%, respectively, in the parental clone, 'Narmada'. The contents of alcohols and their esters (linalool, citronellol, geraniol, citronellyl formate and geranyl formate) in the essential oil of N75 in SC2/VM2 and SC3/VM3 generations were 49% and 35%, respectively, as compared with 69% and 63%, respectively, in the parental clone, 'Narmada'. This is the first report on a chemovariant of rose-scented geranium with a moderately high content of isomenthone. All earlier reported isomenthone-rich variants of rose-scented geranium had quite high contents of isomenthone (64-71%) in their essential oils. The probable modes of origin of this somaclonal variant, its parental clone 'Narmada' (with very low content of isomenthone) and four earlier reported isomenthone-rich variants of Indian cultivars of geranium are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Sigma-Aldrich
Nerol, ≥97%, FCC, FG
Sigma-Aldrich
(R)-(+)-β-Citronellol, 95%
Sigma-Aldrich
(S)-(−)-β-Citronellol, ≥97%
Supelco
(−)-β-Citronellol, analytical standard
Supelco
(±)-β-Citronellol, analytical standard