Merck
  • Substituent effects on zinc phthalocyanine derivatives: a theoretical calculation and screening of sensitizer candidates for dye-sensitized solar cells.
Substituent effects on zinc phthalocyanine derivatives: a theoretical calculation and screening of sensitizer candidates for dye-sensitized solar cells.
Journal of molecular graphics & modelling (2012-10-23)
Linlin Yang, Lianshun Guo, Qianqian Chen, Huafei Sun, Hui Yan, Qinghua Zeng, Xianxi Zhang, Xu Pan, Songyuan Dai
ZUSAMMENFASSUNG
A series of unsymmetrical phthalocyanine sensitizer candidates with different donor and acceptor substituents, namely ZnPcBPh, ZnPcBOPh, ZnPcBtBu, ZnPcBN(Ph)₂, ZnPcBNHPh, ZnPcBNH₂, ZnPcBNHCH₃ and ZnPcBN(CH₃)₂, were designed and calculated using density functional theory (DFT) and time-dependent DFT calculations. The molecular orbital energy levels, the molecular orbital spatial distributions and the electronic absorption spectra of the ZnPcB series molecules were compared with those of TT7 and TT8 to reveal the substituent effects of different donor and acceptor groups on the phthalocyanine compounds and select good sesitizer candidates. The results show that some of these compounds have considerably smaller orbital energy gaps, red-shifted absorption bands and better charge-separated states, causing them to absorb photons in the lower energy region. Several new absorption bands emerge in the 400-600 nm region, which makes it possible for them to become panchromatic sensitizers. This characteristic is superior to the phthalocyanine sensitizers reported previously, including the current record holder, PcS6. The sensitizer candidates screened in the current work are very promising for providing good performance and might even challenge the photon-to-electricity conversion efficiency record of 4.6% for phthalocyanine sensitizers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinkphthalocyanin, Dye content 97 %
Sigma-Aldrich
29H,31H-Phthalocyanin, β-form, 98%