Merck
Sim3 shares some common roles with the histone chaperone Asf1 in fission yeast.
FEBS letters (2012-10-24)
Katsuhiro Tanae, Tomitaka Horiuchi, Takuya Yamakawa, Yuzy Matsuo, Makoto Kawamukai
ZUSAMMENFASSUNG
An H3/H4 histone chaperone, Asf1, plays an essential role in maintaining genomic stability in many species, including fission yeast. Here, we showed that overexpression of a CENP-A chaperone Sim3 suppressed the temperature sensitive phenotype of asf1-33 and asf1-30 mutants and the defect in chromatin structure, and prevented the accumulation of DNA damage in asf1-33 mutants at high temperatures. Furthermore, asf1-33 and Δsim3 were synthetic lethal. Consistent with this, shutdown of sim3 expression in asf1-33 Δsim3 double mutants that contained extragenic sim3 resulted in growth retardation. In addition, the Δsim3 mutant displayed sensitivity to thiabendazol and hydroxyurea, which suggests that Sim3 plays a general role in maintaining chromatin structure. Our results suggest a possibility that Sim3 functions as a histone chaperone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiabendazol, ≥99%, powder
Sigma-Aldrich
Thiabendazol, BioReagent, suitable for plant cell culture, powder
Supelco
Thiabendazol, PESTANAL®, analytical standard
Supelco
Thiabendazol, Pharmaceutical Secondary Standard; Certified Reference Material
Thiabendazol, European Pharmacopoeia (EP) Reference Standard