Merck
Accidental digitoxin intoxication: an interplay between laboratory and clinical medicine.
Biochemia medica (2012-10-25)
Luigi M Castello, Sophie Negro, Francesca Santi, Isabella Zanotti, Matteo Vidali, Marco Bagnati, Giorgio Bellomo, Gian Carlo Avanzi
ZUSAMMENFASSUNG
Two Italian adults arrived at the Emergency Department referring diarrhea, nausea and vomiting for 4 days; weakness, fatigue and visual hallucinations were also complained of. Patients reported the ingestion of some leaves of a plant, which they supposed to be "donkey ears", a week before. Physical examination showed hypotension and bradycardia and ECG examination disclosed sinus rhythm and repolarization abnormalities (scooping of the ST-T complex) in both patients and a 2:1 AV block in the man. Digoxin concentration was evaluated twice for each patient (at the admission and after 4 hours) by the automated immunoassay system ADVIA Centaur. Digitoxin concentration was evaluated by liquid chromatography-mass spectrometry (LC-MS/MS). Despite clinical picture was suggestive of digitalis intoxication, digoxin levels were undetectable. Due to the more severe clinical picture, the male patient was treated with anti-digoxin antibodies (Digifab) achieving a good clinical improvement and remission of the AV block within two hours. Initial diagnosis was confirmed by LC-MS/MS showing high digitoxin concentrations, but digoxin was undetectable. Patients remained stable and 48 hours later were discharged from the hospital. Whereas digoxin determination frequently relies on monoclonal antibodies which do not cross-react to digitoxin, polyclonal antibodies constituting Digifab recognize a large spectrum of cardiac glycosides, including digitoxin. This report emphasizes the primary role of the clinical approach to patients in the emergency setting and how an active communication and a continuous sharing of professional experiences between Laboratory and Clinicians ensure an early and correct diagnosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Digitoxin, European Pharmacopoeia (EP) Reference Standard