Merck
  • Intrinsic reaction-cycle time scale of Na+,K+-ATPase manifests itself in the lipid-protein interactions of nonequilibrium membranes.
Intrinsic reaction-cycle time scale of Na+,K+-ATPase manifests itself in the lipid-protein interactions of nonequilibrium membranes.
Proceedings of the National Academy of Sciences of the United States of America (2012-10-25)
Hélène Bouvrais, Flemming Cornelius, John H Ipsen, Ole G Mouritsen
ZUSAMMENFASSUNG
Interaction between integral membrane proteins and the lipid-bilayer component of biological membranes is expected to mutually influence the proteins and the membrane. We present quantitative evidence of a manifestation of the lipid-protein interactions in liposomal membranes, reconstituted with actively pumping Na(+),K(+)-ATPase, in terms of nonequilibrium shape fluctuations that contain a relaxation time, τ, which is robust and independent of the specific fluctuation modes of the membrane. In the case of pumping Na(+)-ions, analysis of the flicker-noise temporal correlation spectrum of the liposomes leads to τ ~/= 0.5 s, comparing favorably with an intrinsic reaction-cycle time of about 0.4 s from enzymology.