Merck
A novel mouse model of podocyte depletion.
Nephron. Experimental nephrology (2012-10-26)
L Wang, Y Tang, D N Howell, P Ruiz, R F Spurney
ZUSAMMENFASSUNG
The goal of this study was to examine the capacity for glomerular repair after a podocyte-depleting injury. We created transgenic (TG) mice expressing the yeast enzyme cytosine deaminase specifically in glomerular podocytes. In these TG animals, the prodrug 5-flucytosine (5-FC) is converted to 5-fluorouracil and promotes cell death. Treatment with increasing dosages of 5-FC caused graded increases in proteinuria 1-2 weeks after treatment, which returned to control levels by the 10-week time point. Light microscopic examination revealed minimal pathology at the 2-week time point, but electron microscopy revealed found foot process effacement as well as focal areas of glomerular basement membrane duplication, and immunohistochemical studies detected podocyte apoptosis and a decrease in the number of Wilms' tumor protein 1 (WT1)-positive cells. By the 10-week time point, however, the number of WT1-positive cells was similar to controls and a few mice had developed focal areas of glomerulosclerosis. Consistent with the effects of 5-FC on podocyte number, expression of the podocyte mRNAs for nephrin, podocin, synaptopodin and podocalyxin were altered in a similar temporal fashion. The glomerulus has a significant capacity for repair after a podocyte-depleting injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorcytosin, nucleoside analog