- Glutaminolysis feeds mTORC1.
Glutaminolysis feeds mTORC1.
Cell cycle (Georgetown, Tex.) (2012-10-26)
Raúl V Durán, Michael N Hall
PMID23095634
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Ketoglutarsäure, BioReagent, suitable for cell culture, suitable for insect cell culture