Merck
  • Compensation for matrix effects in the gas chromatography-mass spectrometry analysis of 186 pesticides in tea matrices using analyte protectants.
Compensation for matrix effects in the gas chromatography-mass spectrometry analysis of 186 pesticides in tea matrices using analyte protectants.
Journal of chromatography. A (2012-10-30)
Yan Li, Xi Chen, Chunlin Fan, Guofang Pang
ZUSAMMENFASSUNG
A gas chromatography-mass spectrometry (GC-MS) analytical method was developed for simultaneously determining 186 pesticides in tea matrices using analyte protectants to counteract the matrix-induced effect. The matrix effects were evaluated for green, oolong and black tea, representing unfermented, partially fermented and completely fermented teas respectively and depending on the type of tea, 72%, 94% and 94% of the pesticides presented strong response enhancement effect. Several analyte protectants as well as certain combinations of these protectants were evaluated to check their compensation effects. A mixture of triglycerol and d-ribonic acid-γ-lactone (both at 2mg/mL in the injected samples) was found to be the most effective in improving the chromatographic behavior of the 186 pesticides. More than 96% of the 186 pesticides achieved recoveries within the range of 70-120% when using the selected mixture of analyte protectants. The simple addition of analyte protectants offers a more convenient solution to overcome matrix effects, results in less active sites compared to matrix-matched standardization and can be an effective approach to compensate for matrix effects in the GC-MS analysis of pesticide residues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
Triglycerin, technical
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture