Merck
  • Azide-alkyne cycloaddition en route towards 1H-1,2,3-triazole-tethered β-lactam-ferrocene and β-lactam-ferrocenylchalcone conjugates: synthesis and in vitro anti-tubercular evaluation.
Azide-alkyne cycloaddition en route towards 1H-1,2,3-triazole-tethered β-lactam-ferrocene and β-lactam-ferrocenylchalcone conjugates: synthesis and in vitro anti-tubercular evaluation.
Dalton transactions (Cambridge, England : 2003) (2012-10-31)
Kewal Kumar, Séverine Carrère-Kremer, Laurent Kremer, Yann Guérardel, Christophe Biot, Vipan Kumar
ZUSAMMENFASSUNG
A diverse range of triazoles were prepared following well established, Cu-mediated azide-alkyne cycloaddition reactions with the aim of probing the anti-tubercular structure-activity relationships (SAR) within the β-lactam-ferrocene-triazole conjugate family. The anti-tubercular evaluation studies of the synthesized conjugates revealed that none of the scaffolds exhibited any activity that restricted mycobacterial growth even at high doses. The introduction of various substituents onto the N-1 of the β-lactam ring, introducing mono- or di-ferrocenylchalcone substituents at the C-3 position as well as introducing a spacer of varying chain length failed to produce any significant enhancement in the activity profiles. The described protocol was a successful attempt on the inclusion of a ferrocene nucleus in the β-lactam family tethered via triazole linkers having metabolic stability and physicochemical favourability.