Merck
Ergotamine-associated valvulopathy with recurrent chylous pleural effusion.
Texas Heart Institute journal (2012-10-31)
John P Bois, Vara Venkata Ponnada, D Fearghas O'Cochlain, Peter A Brady, William D Edwards, Kyle W Klarich
ZUSAMMENFASSUNG
We report a rare case of ergotamine-associated mitral stenosis in a 55-year-old woman who presented with recurrent chylous pleural effusion. Echocardiographic, gross, and microscopic features of the mitral valve were consistent with chronic ergotamine-induced valvulopathy. We conclude that medication-induced valvulopathy should be included in the differential diagnosis of valvular heart disease. In addition, cardiac function should be monitored before and during long-term therapy with ergotamine or ergotamine-derived dopamine agonists.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ergotamin -D-tartrat, ≥97.0% (calc. based on dry substance, NT)