- Metric similarity of dynamic commutation processes in situ and in vitro.
Metric similarity of dynamic commutation processes in situ and in vitro.
Bulletin of experimental biology and medicine (2012-11-01)
S V Stovbun, A A Skoblin, A M Zanin, A I Mikhailov, V A Tverdislov, E E Bragina, Yu M Rybin, I M Ageev, G G Shishkin
PMID23113299
ZUSAMMENFASSUNG
The dynamics of string growth was studied in model homochiral solutions of biomimetics, trifluoroacetylated amino alcohols (TFAAA) in heptane, water, and inverted heptane-water emulsion. In heptane and water, a thick (~1 μ in diameter) string had a crown of thin strings on its growing terminal and these thin strings effectively adsorbed dissolved TFAAA. In emulsion, the strings grew inside the water droplets, in which this TFAAA cannot be solved, presumably due to transport of TFAAA molecules from heptane into water in the surface layer surrounding the string. Applications of these phenomena to in vivo cell commutation were discussed.