Merck
  • Regulation of Salmonella flagellin-induced interleukin-8 in intestinal epithelial cells by muramyl dipeptide.
Regulation of Salmonella flagellin-induced interleukin-8 in intestinal epithelial cells by muramyl dipeptide.
Cellular immunology (2012-11-06)
Fu-Chen Huang
ZUSAMMENFASSUNG
Toll-like receptor 5 (TLR5) and nucleotide-binding oligomerization domain 2 (Nod2) are two important pattern recognition receptors involved in innate immunity to invading pathogens. Flagellin, recognized by TLR5, is Salmonella's dominant pro-inflammatory determinant in intestinal epithelial cells (IECs). Nod2 has played a pivotal role in protecting against intestinal bacterial infection. Therefore the aim of the study is to investigate regulation of Salmonella flagellin-induced interleukin (IL)-8 (IL-8) in IECs by Nod2 agonist, muramyl dipeptide (MDP). We found that MDP by itself induced only a weak IL-8 secretion in Caco-2 cells. However, it did show synergistic enhancement on flagellin-induced IL-8 production in Caco-2 cells, possibly caused by flagellin-mediated enhanced Nod2 recruitment into cell membrane. By Western blot and siRNA, we showed ERK and NF-κB, Nod2 and Rip2 were involved in the synergistic effect of MDP. These findings suggested that the cooperation of TLR5 and Nod2 in IECs regulates inflammatory response to Salmonella infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetylmuramyl-L-alanyl-D-isoglutamin Hydrat, ≥98% (TLC)