Merck
  • Encapsulation of food grade antioxidant in natural biopolymer by electrospinning technique: a physicochemical study based on zein-gallic acid system.
Encapsulation of food grade antioxidant in natural biopolymer by electrospinning technique: a physicochemical study based on zein-gallic acid system.
Food chemistry (2012-11-06)
Yun Ping Neo, Sudip Ray, Jianyong Jin, Marija Gizdavic-Nikolaidis, Michel K Nieuwoudt, Dongyan Liu, Siew Young Quek
ZUSAMMENFASSUNG
Gallic acid was successfully incorporated into zein ultra-fine fibres at different loading amount (5%, 10% and 20%) in order to develop an encapsulating technology for functional ingredient delivery using electrospinning. The produced fibres exhibit diameters ranging from 327 to 387 nm. The physical and thermal properties of encapsulated gallic acid were determined by X-ray diffraction (XRD) and differential scanning calorimetry (DSC); and the interaction between gallic acid and zein was attested by attenuated total reflection-Fourier transform infrared (ATR-FTIR). Thermogravimetric analysis (TGA) demonstrated a different thermal stability of the fabricated complex before and after the gallic acid incorporation. Lastly, the 1,1'-diphenyl-2-picrylhydrazyl (DPPH) assay showed that the gallic acid had retained its antioxidant activity after incorporation in zein electrospun fibres. Overall, electrospinning technique had shown promising results as an efficient and effective method for the preparation of sub-micron structured encapsulated functional ingredient that may find uses in food industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zein aus Mais