Merck
  • Design, synthesis and antibacterial activities of vanillic acylhydrazone derivatives as potential β-ketoacyl-acyl carrier protein synthase III (FabH) inhibitors.
Design, synthesis and antibacterial activities of vanillic acylhydrazone derivatives as potential β-ketoacyl-acyl carrier protein synthase III (FabH) inhibitors.
European journal of medicinal chemistry (2012-11-06)
Xiao-Liang Wang, Yan-Bin Zhang, Jian-Feng Tang, Yu-Shun Yang, Ruo-Qi Chen, Fei Zhang, Hai-Liang Zhu
ZUSAMMENFASSUNG
Fatty acid biosynthesis is essential for bacterial survival. FabH, β-ketoacyl-acyl carrier protein (ACP) synthase III, is a particularly attractive target, since it is central to the initiation of fatty acid biosynthesis and is highly conserved among Gram-positive and Gram-negative bacteria. A series of acylhydrazone derivatives were synthesized and developed as potent inhibitors of FabH. This inhibitor class demonstrates strong broad-spectrum antibacterial activity. Compounds with potent antibacterial activities were tested for their Escherichia coli FabH inhibitory activity. Especially, compound E9 showed the most potent antibacterial activity with MIC values of 0.39-1.56 μg/mL against the tested bacterial strains and exhibited the most potent E. coli FabH inhibitory activity with IC(50) of 2.5 μM. Docking simulation was performed to position compound E9 into the E. coli FabH active site to determine the probable binding conformation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vanillinsäure, 97%
Sigma-Aldrich
Vanillinsäure, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillinsäure, ≥97%, FG
Supelco
Vanillinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland