Merck
  • CdO nanosheet film with a (200)-preferred orientation with sensitivity to liquefied petroleum gas (LPG) at low-temperatures.
CdO nanosheet film with a (200)-preferred orientation with sensitivity to liquefied petroleum gas (LPG) at low-temperatures.
Physical chemistry chemical physics : PCCP (2012-11-08)
Guangliang Cui, Zimeng Li, Liang Gao, Mingzhe Zhang
ZUSAMMENFASSUNG
CdO nanosheet film can be synthesized by electrochemical deposition in an ultra-thin liquid layer by using Cd(NO(3))(2) and HNO(3) as source materials for Cd and oxygen respectively. HNO(3) is also used to adjust the pH of the electrolyte. Studies on the detailed structure indicate that the synthesized CdO nanosheet film has a face-centered cubic structure with (200)-preferred orientation. The response of the CdO nanosheet film to liquefied petroleum gas (LPG) at low temperature has been significantly improved by the novel structure of film. It has exhibited excellent sensitivity and selectivity to LPG at low temperature. A new growth mechanism of electrochemical deposition has been proposed to elaborate the formation of nanosheet in an ultra-thin liquid layer. The self-oscillation of potential in the growth interface and intermediate hydroxide are responsible for the formation of nanosheets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmiumoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmiumoxid, powder, 99.5% trace metals basis