Merck
  • Construction of an artificial pathway for isobutanol biosynthesis in the cytosol of Saccharomyces cerevisiae.
Construction of an artificial pathway for isobutanol biosynthesis in the cytosol of Saccharomyces cerevisiae.
Bioscience, biotechnology, and biochemistry (2012-11-08)
Fumio Matsuda, Takashi Kondo, Kengo Ida, Hironori Tezuka, Jun Ishii, Akihiko Kondo
ZUSAMMENFASSUNG
To increase isobutanol production in Saccharomyces cerevisiae, the valine biosynthetic pathway was activated by overexpression of the relevant enzymes in the mitochondria and the cytosol. Native mitochondrial enzymes were overepxressed in the cytosol by deleting the mitochondrial transit peptides. The metabolically engineered S. cerevisiae possessing the cytosolic pathway showed increased isobutanol production (63 ± 4 mg/L).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Methyl-1-propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, ACS reagent, ≥99.0%
Sigma-Aldrich
Isobutylalkohol, ≥99%, FCC, FG
Sigma-Aldrich
2-Methyl-1-propanol, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, anhydrous, 99.5%
Supelco
2-Methyl-1-propanol, analytical standard
Sigma-Aldrich
Isobutylalkohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
2-Methyl-1-propanol, BioUltra, Molecular Biology, ≥99.5% (GC)