Merck
FeCl3-catalyzed cyclization of α-sulfonamido-allenes with aldehydes--the substituent effect.
Chemical communications (Cambridge, England) (2012-11-09)
Jiajia Cheng, Xinjun Tang, Yihua Yu, Shengming Ma
ZUSAMMENFASSUNG
FeCl(3)-catalyzed aza-Prins-cyclization reaction of α-sulfonamido-allenes with aldehydes afforded 1,2,3,6-tetrahydropyridine or 2,5-dihydro-1H-pyrrole derivatives efficiently and highly selectively. The different regioselectivity is probably caused by the stabilizing effect of the phenyl group on the positively charged allylic intermediate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eisen(III)-chlorid Hexahydrat, ACS reagent, 97%
Sigma-Aldrich
Eisen(III)-chlorid Hexahydrat, reagent grade, ≥98%, chunks
Sigma-Aldrich
Eisen(III)-chlorid Hexahydrat, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Eisen(III)-chlorid, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Eisen(III)-chlorid -Lösung, purum, 45% FeCl3 basis
Sigma-Aldrich
Eisen(III)-chlorid, ≥99.9% trace metals basis
Sigma-Aldrich
Eisen(III)-chlorid, 0.2 M in 2-methyltetrahydrofuran
Millipore
TDA-Reagenz, suitable for microbiology