Merck
5-Hydroxyindoles by intramolecular alkynol-furan diels-alder cycloaddition.
The Journal of organic chemistry (2012-11-10)
Matthew LaPorte, Ki Bum Hong, Jie Xu, Peter Wipf
ZUSAMMENFASSUNG
A convergent approach provides a convenient access to synthetically and biologically useful 3,4-disubstituted 5-hydroxyindoles. The one-pot procedure uses microwave heating to initiate an intramolecular [4 + 2]-cycloaddition of an alkynol segment onto a furan followed by a fragmentation, aromatization, and N-Boc deprotection cascade. Yields range from 15 to 74%, with aromatic substituents providing better conversions. 4-Trimethylsilylated analogues undergo a 1,3-silatropic rearrangement to give the O-TMS ethers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
5-Hydroxy-indol, 97%
Supelco
Furan, analytical standard