Merck
  • A vitamin D3 analog augmented interleukin-8 production by human monocytic cells in response to various microbe-related synthetic ligands, especially NOD2 agonistic muramyldipeptide.
A vitamin D3 analog augmented interleukin-8 production by human monocytic cells in response to various microbe-related synthetic ligands, especially NOD2 agonistic muramyldipeptide.
International immunopharmacology (2012-11-20)
Tomoko Ikeuchi, Takashi Nakamura, Satoshi Fukumoto, Haruhiko Takada
ZUSAMMENFASSUNG
Active metabolite vitamin D(3), 1α,25-dihydroxyvitamin D(3), is a pleiotropic factor and exhibits various physiological functions, including immunomodulating activities. In this study, the possible regulation of innate immune responses of human monocytic cells by a vitamin D(3) analog was examined. Human monocytic THP-1 cells were pre-treated with OCT, vitamin D(3) analog, 1α,25-dihydroxy-22-oxavitamin D(3), followed by stimulation with various chemically synthesized Toll-like receptors (TLR) and NOD1 and NOD2 ligands. OCT-treated cells produced more IL-8 than non-treated cells upon stimulation with various chemically-synthesized ligands: TLR2-agonistic lipopeptide (FSL-1), TLR3-agonistic poly I:C, TLR4-agonistic lipid A (E. coli-type LA-15-PP), NOD1-agonistic FK565 and NOD2-agonistic muramyldipeptide (MDP). Among the ligands, MDP was the highest inducer of IL-8 production in OCT-treated THP-1 cells, and IL-8 production increased depending on the treatment time until 72h. OCT up-regulated the expression of NOD2 in THP-1 cells, and OCT-treated cells exhibited higher activation of p38, JNK and ERK in the MAPK pathway, IκBα in the NF-κB pathway, and TAK1 upstream in response to MDP than non-treated cells. Analysis using siRNA against NOD2 and inhibitors of specific signal molecules indicated that the existence of NOD2 and activation of the above signaling molecules are required for enhanced production of IL-8 in OCT-treated THP-1 cells. These findings suggested that NOD2, NF-κB and MAPK pathways are involved in the activity of OCT to augment the response of human monocytic cells to MDP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetylmuramyl-L-alanyl-D-isoglutamin Hydrat, ≥98% (TLC)