Merck
  • Hydrophobic interaction-mediated capture and release of cancer cells on thermoresponsive nanostructured surfaces.
Hydrophobic interaction-mediated capture and release of cancer cells on thermoresponsive nanostructured surfaces.
Advanced materials (Deerfield Beach, Fla.) (2012-11-20)
Hongliang Liu, Xueli Liu, Jingxin Meng, Pengchao Zhang, Gao Yang, Bin Su, Kang Sun, Li Chen, Dong Han, Shutao Wang, Lei Jiang
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(N-isopropylacrylamid), Mn ~40,000
Sigma-Aldrich
Poly(N-Isopropylacrylamid), Carboxylsäure-Endgruppe, average Mn 10,000
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000