Merck
eComment. The problem of anaemia correction in cardiac surgery patients.
Interactive cardiovascular and thoracic surgery (2012-11-21)
Leo A Bockeria, Olga Bockeria, Maria Sokolskaya, Sergei Donakanian