Merck
  • Arylation of 2-substituted pyridines via Pd-catalyzed decarboxylative cross-coupling reactions of 2-picolinic acid.
Arylation of 2-substituted pyridines via Pd-catalyzed decarboxylative cross-coupling reactions of 2-picolinic acid.
Chemical communications (Cambridge, England) (2012-11-23)
Xinjian Li, Dapeng Zou, Faqiang Leng, Chunxia Sun, Jingya Li, Yangjie Wu, Yusheng Wu
ZUSAMMENFASSUNG
The novel palladium-catalyzed decarboxylative cross-coupling reactions of 2-picolinic acid with aryl and heteroaryl bromides including benzenes, naphthalenes, pyridines and quinolines for C-C bond formation have been successfully achieved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
2-Picolinsäure, ReagentPlus®, 99%
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Supelco
Naphthalin -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland