Merck
  • [Construction of ompW knock-out mutants of Escherichia coli to increase sensitivity to neomycinsulphate and ampicillin].
[Construction of ompW knock-out mutants of Escherichia coli to increase sensitivity to neomycinsulphate and ampicillin].
Wei sheng wu xue bao = Acta microbiologica Sinica (2012-11-24)
Xianbin Wu, Haijie Zou, Lihua Tian, Jianyi Pan, Fukun Zhao
ZUSAMMENFASSUNG
To investigate the contribution of an outer membrane protein OmpW to tolerance neomycinsulphate and ampicillin of Escherichia coli K12. The ompW knock-out mutant (deltaompW) of E. coli K12 was generated using lambda-Red recombination system. Then the minimal inhibitory concentration (MIC) and the survival rates under 1/2 MIC of neomycinsulphate or ampicillin of deltaompW and E. coli K12 were determined respectively. The deltaompW was successfully obtained through confirmation of PCR analysis at the gene level and Western blot analysis at the protein level. The MIC of neomycinsulphate of deltaompW is 1.7 microg/mL. The value is much lower than that of E. coli K12, which is 8.0 microg/mL. Difference of survival rates under 1/2 MIC of neomycinsulphate of deltaompW and E. coli K12 was also observed, and their survival rates are 39% and 98% , respectively. The MIC of ampicillin of deltaompW is 3.3 microg/mL. The value is also lower than that of E. coli K12 (16.0 microg/mL). The survival rates under 1/2 MIC ampicillin of deltaompW and E. coli K12 are 30.3% and 70.38%, respectively. The AompW is much more sensitive to neomycinsulphate and ampicillin than its parent strain. The result indicated that OmpW played crucial role in bacteria resistance of drug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Neomycin -trisulfat (Salz) Hydrat, powder
Sigma-Aldrich
Neomycin -trisulfat (Salz) Hydrat, powder, BioReagent, suitable for cell culture
Supelco
Neomycin -trisulfat (Salz) Hydrat, VETRANAL®, analytical standard
Sigma-Aldrich
Neomycin -trisulfat (Salz) Hydrat, meets USP testing specifications, powder
Neomycin-Sulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Neomycin -trisulfat (Salz) Hydrat, suitable for plant cell culture