Merck
Mn2+ modulates the kinetic properties of an archaeal member of the PLL family.
Chemico-biological interactions (2012-11-24)
Elena Porzio, Spartaco Di Gennaro, Achille Palma, Giuseppe Manco
ZUSAMMENFASSUNG
Recently we reported on the characterization of an archaeal member of the amidohydrolase superfamily, namely Sulfolobus acidocaldarius lactonase, showing low but significant and extremely thermostable paraoxonase activity. This enzyme, that we have named SacPox, is a member of the new described family of phosphotriesterase-like lactonases (PLLs). In this family the binuclear metal centre, which is involved in the catalytic machinery, has been poorly studied up to now. In this work we describe the expression of the protein in presence of different metals showing Mn(2+) to support the higher activity. The enzyme has been over-expressed, purified and characterized as a Mn(2+)-containing enzyme by inductive plasma coupled mass spectrometry (ICP-MS), showing also surprising kinetic differences in comparison with the cadmium-containing enzyme. The Mn(2+) containing enzyme was about 30-fold more efficient with paraoxon as substrate and more stable than the Cd(2+) counterpart, even though the Mn(2+) affinity for the binuclear metal centre is apparently lower. These results increase our knowledge of the biochemical characteristics of SacPox mainly with regard to the metal-ions modulation of function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paraoxon-Ethyl, ≥90%, oil
Supelco
Paraoxonethyl, PESTANAL®, analytical standard