Merck
  • Development and application of salting-out assisted liquid/liquid extraction for multi-mycotoxin biomarkers analysis in pig urine with high performance liquid chromatography/tandem mass spectrometry.
Development and application of salting-out assisted liquid/liquid extraction for multi-mycotoxin biomarkers analysis in pig urine with high performance liquid chromatography/tandem mass spectrometry.
Journal of chromatography. A (2012-11-28)
Suquan Song, Emmanuel Njumbe Ediage, Aibo Wu, Sarah De Saeger
ZUSAMMENFASSUNG
Direct determination of urinary mycotoxins is a better approach to assess individual's exposure than the indirect estimation from average dietary intakes. In this study, a new analytical method was developed and validated for simultaneous analysis of aflatoxin B1, deoxynivalenol, fumonisin B1, ochratoxin A, zearalenone and T2 toxin and their metabolites in pig urine. In total 12 analytes were selected. A salting-out assisted liquid-liquid extraction procedure was used for sample preparation. High performance liquid chromatography/tandem mass spectrometry was used for the separation and detection of all the analytes. The extraction recoveries were in a range of 70-108%, with the intra-day relative standard deviation and inter-day relative standard deviation lower than 25% for most of the compounds at 3 different concentration levels. Meanwhile the method bias for all the analytes did not exceed 20%. The limits of quantification ranged from 0.07ngmL(-1) for ochratoxin A to 3.3ngmL(-1) for deoxynivalenol. Matrix effect was evaluated in this study and matrix-matched calibration was used for quantification. The developed method was also validated for human urine as an extension of its application. Finally, the developed method was applied in a pilot study to analyze 28 pig urine samples. Deoxynivalenol, aflatoxin B1, fumonisin B1 and ochratoxin A were detected in these samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aflatoxin B1, from Aspergillus flavus
Sigma-Aldrich
Fumonisin B1 aus Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Zearalenon, fungal mycotoxin
Supelco
Aflatoxin B1 -Lösung, 2 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
T-2-Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Supelco
Zearalenon -Lösung, 100 μg/mL in acetonitrile, analytical standard
Supelco
T2-Toxin -Lösung, 100 μg/mL in acetonitrile, analytical standard
Supelco
Fumonisin B1 -Lösung, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Aflatoxin B1 -Lösung, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 -Lösung, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard