Merck
  • Ultrasonic-assisted ozone oxidation process of triphenylmethane dye degradation: evidence for the promotion effects of ultrasonic on malachite green decolorization and degradation mechanism.
Ultrasonic-assisted ozone oxidation process of triphenylmethane dye degradation: evidence for the promotion effects of ultrasonic on malachite green decolorization and degradation mechanism.
Bioresource technology (2012-12-04)
Xian-Jiao Zhou, Wan-Qian Guo, Shan-Shan Yang, He-Shan Zheng, Nan-Qi Ren
ZUSAMMENFASSUNG
This study aimed to prove the promotion effects of ultrasonic on malachite green (MG) decolorization in the ultrasonic-assisted ozone oxidation process (UAOOP), and propose the possible pathway of MG degradation. The decolorization of MG followed an apparent pseudo first-order kinetic law (initial MG concentration 100-1000 mg/L). When ultrasonic (US) was applied with ozone simultaneously, the apparent pseudo-first-order rate constant (K(app)) increased, and the time MG decolorized to the half of initial concentration (T(1/2)) shortened 185 s (1000 mg/L). Moreover, the stoichiometric ratio (Z(app)) between O(3) and MG was enhanced by US to 2.0 mol, saving 11% oxidant addition, comparing to individual ozone process. These results indicated that the application of US can reduce reaction time and dose of ozone addition. The possible pathway of MG degradation included three major approaches. And the result suggested that the reaction between MG and hydroxyl radical was substitution reaction rather than adduct reaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Malachitgrün Oxalat, certified by the BSC
Sigma-Aldrich
Malachitgrün Oxalat, suitable for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Triphenylmethan, 99%
Sigma-Aldrich
Malachitgrün Oxalat, Technical grade
Sigma-Aldrich
Malachitgrün -Lösung, suitable for microscopy
Supelco
Malachitgrün Oxalat, VETRANAL®, analytical standard
Sigma-Aldrich
Schaeffer und Fulton-Sporen-Färbelösung A, suitable for microscopy