Merck
  • Highly sensitive ochratoxin A impedimetric aptasensor based on the immobilization of azido-aptamer onto electrografted binary film via click chemistry.
Highly sensitive ochratoxin A impedimetric aptasensor based on the immobilization of azido-aptamer onto electrografted binary film via click chemistry.
Talanta (2012-12-04)
Akhtar Hayat, Audrey Sassolas, Jean-Louis Marty, Abd-Elgawad Radi
ZUSAMMENFASSUNG
The aptamer immobilization onto organized mixed layers of diazonium salts via click chemistry was explored. The immobilized aptamer was employed in the fabrication of a highly sensitive and reusable electrochemical impedimetric aptasensor for the detection of ochratoxin A (OTA). The screen-printed carbon electrodes (SPCEs) were first modified by electrografting of a protected 4-((trimethylsilyl)ethynyl) benzene (TMSi-Eth-Ar) layer followed by a second one of p-nitrobenzene (p-NO(2)-Ar) by means of electrochemical reduction of their corresponding diazonium salts, (TMSi-Eth-Ar-N(2)(+)) and (p-NO(2)-ArN(2)(+)). After deprotection, a layer with active ethynyl groups was obtained. In the presence of copper (I) catalyst, the ethynyl groups reacted efficiently with aptamer bearing an azide function, thus forming a covalent 1,2,3-triazole linkage. Cyclic voltammetry (CV) and electrochemical impedance spectroscopy (EIS) in the presence of ferri/ferrocyanide redox probe [Fe(CN)(6)](4-/3-) were used to characterize each step in the aptasensor development. The increase in electron-transfer resistance (R(et)) values due to the specific aptamer-OTA interaction was proportional to the concentration of OTA in a range between 1.25 ng/L and 500 ng/L, with a detection limit of 0.25 ng/L.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ochratoxin A -Lösung, 10 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Ochratoxin A, from Petromyces albertensis, ≥98% (HPLC)