Merck
Increased dietary intake of vitamin A promotes aortic valve calcification in vivo.
Arteriosclerosis, thrombosis, and vascular biology (2012-12-04)
Danielle J Huk, Harriet L Hammond, Hiroyuki Kegechika, Joy Lincoln
ZUSAMMENFASSUNG
Calcific aortic valve disease (CAVD) is a major public health problem with no effective treatment available other than surgery. We previously showed that mature heart valves calcify in response to retinoic acid (RA) treatment through downregulation of the SRY transcription factor Sox9. In this study, we investigated the effects of excess vitamin A and its metabolite RA on heart valve structure and function in vivo and examined the molecular mechanisms of RA signaling during the calcification process in vitro. Using a combination of approaches, we defined calcific aortic valve disease pathogenesis in mice fed 200 IU/g and 20 IU/g of retinyl palmitate for 12 months at molecular, cellular, and functional levels. We show that mice fed excess vitamin A develop aortic valve stenosis and leaflet calcification associated with increased expression of osteogenic genes and decreased expression of cartilaginous markers. Using a pharmacological approach, we show that RA-mediated Sox9 repression and calcification is regulated by classical RA signaling and requires both RA and retinoid X receptors. Our studies demonstrate that excess vitamin A dietary intake promotes heart valve calcification in vivo. Therefore suggesting that hypervitaminosis A could serve as a new risk factor of calcific aortic valve disease in the human population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Retinylpalmitat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinylpalmitat, potency: ≥1,700,000 USP units per g
Supelco
Retinolpalmitat, analytical standard
Sigma-Aldrich
Retinylpalmitat, Type IV, ~1,800,000 USP units/g, oil