Merck
  • Ex situ treatment of N-nitrosodimethylamine (NDMA) in groundwater using a fluidized bed reactor.
Ex situ treatment of N-nitrosodimethylamine (NDMA) in groundwater using a fluidized bed reactor.
Water research (2012-12-05)
Todd S Webster, Charles Condee, Paul B Hatzinger
ZUSAMMENFASSUNG
N-nitrosodimethylamine (NDMA) is a suspected human carcinogen that has traditionally been treated in water using ultraviolet irradiation (UV). The objective of this research was to examine the application of a laboratory-scale fluidized bed reactor (FBR) as an alternative technology for treating NDMA to part-per-trillion (ng/L) concentrations in groundwater. Previous studies have shown that the bacterium Rhodococcus ruber ENV425 is capable of cometabolizing NDMA during growth on propane as a primary substrate in batch culture (Fournier et al., 2009) and in a bench-scale membrane bioreactor (Hatzinger et al., 2011) to low ng/L concentrations. R. ruber ENV425 was inoculated into the FBR during this study. With a hydraulic residence time (HRT) of 20 min, the FBR was found to be an effective means to treat 10-20 μg/L of NDMA to effluent concentrations less than 100 ng/L. When the HRT was increased to 30 min and oxygen and propane addition rates were optimized, the FBR system demonstrated treatment of the NDMA to effluent concentrations of less than 10 ng/L. Short-term shutdowns and the presence of trichloroethene (TCE) at 6 μg/L as a co-contaminant had minimal effect on the treatment of NDMA in the FBR. The data suggest that the FBR technology can be a viable alternative to UV for removing NDMA from groundwater.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trichlorethylen, ACS reagent, ≥99.5%
Supelco
N-Nitrosodimethylamin (NDMA) -Lösung, certified reference material, 200 μg/mL in methanol
Supelco
Trichlorethylen, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichlorethylen, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichlorethylen -Lösung, certified reference material, 5000 μg/mL in methanol