Merck
  • New TFA-free cleavage and final deprotection in fmoc solid-phase peptide synthesis: dilute HCl in fluoro alcohol.
New TFA-free cleavage and final deprotection in fmoc solid-phase peptide synthesis: dilute HCl in fluoro alcohol.
Organic letters (2012-12-06)
Pasquale Palladino, Dmitry A Stetsenko
ZUSAMMENFASSUNG
A novel method for cleaving from resin and removing acid-labile protecting groups for the Fmoc solid-phase peptide synthesis is described. 0.1 N HCl in hexafluoroisopropanol or trifluoroethanol cleanly and rapidly removes the tert-butyl ester and ether, Boc, trityl, and Pbf groups and cleaves the common resin linkers: Wang, HMPA, Rink amide, and PAL. Addition of just 5-10% of a hydrogen-bonding solvent considerably retards or even fully inhibits the reaction. However, a non-hydrogen-bonding solvent is tolerated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Supelco
Trifluoressigsäure, analytical standard