Merck
Tyrosyl DNA phosphodiesterase 2, an enzyme fit for purpose.
Nature structural & molecular biology (2012-12-06)
Keith W Caldecott
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
O-Phospho-L-Tyrosin