Merck
  • Preparation and valuation of a topical solution containing eutectic mixture of itraconazole and phenol.
Preparation and valuation of a topical solution containing eutectic mixture of itraconazole and phenol.
Archives of pharmacal research (2012-12-06)
Chun-Woong Park, Ju-Young Kim, Yun-Seok Rhee, Tack-Oon Oh, Jung-Myung Ha, Na-Young Choi, Sang-Cheol Chi, Eun-Seok Park
ZUSAMMENFASSUNG
The purposes of this study were to prepare a topical solution containing itraconazole (ITR)-phenol eutectic mixture and to evaluate its ex vivo skin permeation, in vivo deposition and in vivo irritation. The eutectic mixture was prepared by agitating ITR and phenol (at a weight ratio of 1:1) together at room temperature. The effects of additives on the skin permeation of ITR were evaluated using excised hairless mouse skin. The in vivo skin deposition and skin irritation studies were performed in Sprague-Dawley rat and New Zealand white rabbit model. The permeability coefficient of ITR increased with addition of oleic acid in the topical solution. Otherwise, the permeability coefficient was inversely proportional to the concentration of the thickening agent, HPMC. The optimized topical solution contained 9 wt% of the ITR-phenol eutectic mixture, 9.0 wt% of oleic acid, 5.4 wt% of hydroxypropylmethyl cellulose and 76.6 wt% of benzyl alcohol. The steady-state flux and permeability coefficient of the optimized topical solution were 0.90 ± 0.20 μg/cm(2)·h and 22.73 ± 5.73 × 10(6) cm/h, respectively. The accumulated of ITR in the epidermis and dermis at 12 h was 49.83 ± 9.02 μg/cm(2). The topical solution did not cause irritation to the skins of New Zealand white rabbits. Therefore, the findings of this study indicate the possibilities for the topical application of ITR via an external preparation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzylalkohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzylalkohol, ACS reagent, ≥99.0%
Supelco
Benzylalkohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
Benzylalkohol, anhydrous, 99.8%
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Benzylalkohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Benzylalkohol, ≥99%, FCC, FG
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Supelco
Benzylalkohol, analytical standard
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
Sigma-Aldrich
Benzylalkohol, natural, ≥98%, FG
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000
Benzylalkohol, European Pharmacopoeia (EP) Reference Standard