Merck
Divergent synthesis of trans-fused polycyclic ethers by a convergent oxiranyl anion strategy.
The Journal of organic chemistry (2012-12-12)
Takeo Sakai, Ai Sugimoto, Hiroki Tatematsu, Yuji Mori
ZUSAMMENFASSUNG
Octacyclic polyethers that correspond to the CDEFGHIJ-ring system of yessotoxin as well as G- and/or I-ring-modified analogues were synthesized in a divergent manner, starting from a common intermediate, using an [X + 2 + Y]-type convergent method. Reaction of a triflate with the oxiranyl anion generated from an epoxy sulfone, followed by ring expansion, allowed for the incorporation of medium-sized ring ethers into the key intermediate. Subsequent acetal formation and reductive etherification afforded various octacycles containing seven- and eight-membered ether rings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ethylenoxid -Lösung, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
Ethylenoxid -Lösung, 2.5-3.3 M in THF
Sigma-Aldrich
Ethylenoxid, ≥99.5%