Merck
  • Meta-analysis of the effect of immunocastration on production performance, reproductive organs and boar taint compounds in pigs.
Meta-analysis of the effect of immunocastration on production performance, reproductive organs and boar taint compounds in pigs.
Animal : an international journal of animal bioscience (2012-12-12)
N Batorek, M Čandek-Potokar, M Bonneau, J Van Milgen
ZUSAMMENFASSUNG
Meta-analytical approach was used to quantitatively synthesize the effect of immunocastration on growth, carcass, meat quality, reproductive organs and boar taint compounds. Altogether, 41 papers were collected for effect size (θ) calculation and the comparisons were made with entire males (EM) and surgical castrates (SC). The data for reproductive organs and growth performance are numerous enough to draw firm conclusions. In contrast, data for carcass and meat quality are more limited. Results of meta-analysis show efficient immunocastration with the magnitude of the response being by far the largest for reproductive organs (θ = -2.8 to -5.0) and boar taint substances (θ = -2.8 and -0.8 for androstenone and skatole, respectively). However, compared with SC, the immunocastrates exhibit larger bulbourethral glands (θ = 1.3) and slightly higher concentrations of androstenone and skatole (θ = 0.1 and θ = 0.2, respectively). The impact of immunocastration is also remarkable on performance, where the main advantage of the immunocastrates is their boar-like performance until revaccination. In the period following the second vaccination, they eat much more than EM (θ = 2.1), resulting in large effect size for growth rate compared with both EM and SC (θ = 1.1 and θ = 1.4, respectively). Considering the whole fattening period, their feed conversion ratio is higher compared with EM (θ = 0.6) and much lower than that of SC (θ = -1.3), although exhibiting moderately faster growth compared with both (θ = 0.6 and θ = 0.2, respectively). With regard to carcass quality, the immunocastrates take intermediate position between EM and SC. Besides, our analysis suggests no difference in meat quality with SC and some meat quality advantages of immunocastrates over EM because of higher intramuscular fat content (θ = 0.4) and lower shear force (θ = -0.6).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Methylindol, 98%
Sigma-Aldrich
Skatol, ≥98%
Supelco
cis-Androsteron, VETRANAL®, analytical standard