Merck
  • Measurements of water content in hydroxypropyl-methyl-cellulose based hydrogels via texture analysis.
Measurements of water content in hydroxypropyl-methyl-cellulose based hydrogels via texture analysis.
Carbohydrate polymers (2012-12-12)
Gaetano Lamberti, Sara Cascone, Maria Margherita Cafaro, Giuseppe Titomanlio, Matteo d'Amore, Anna Angela Barba
ZUSAMMENFASSUNG
In this work, a fast and accurate method to evaluate the water content in a cellulose derivative-based matrix subjected to controlled hydration was proposed and tuned. The method is based on the evaluation of the work of penetration required in the needle compression test. The work of penetration was successfully related to the hydrogel water content, assayed by a gravimetric technique. Moreover, a fitting model was proposed to correlate the two variables (the water content and the work of penetration). The availability of a reliable tool is useful both in the quantification of the water uptake phenomena, both in the management of the testing processes of novel pharmaceutical solid dosage forms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000