Merck
Taming of fluoroform: direct nucleophilic trifluoromethylation of Si, B, S, and C centers.
Science (New York, N.Y.) (2012-12-12)
G K Surya Prakash, Parag V Jog, Patrice T D Batamack, George A Olah
ZUSAMMENFASSUNG
Fluoroform (CF(3)H), a large-volume by-product of the manufacture of Teflon, refrigerants, polyvinylidene fluoride (PVDF), fire-extinguishing agents, and foams, is a potent and stable greenhouse gas that has found little practical use despite the growing importance of trifluoromethyl (CF3) functionality in more structurally elaborate pharmaceuticals, agrochemicals, and materials. Direct nucleophilic trifluoromethylation using CF(3)H has been a challenge. Here, we report on a direct trifluoromethylation protocol using close to stoichiometric amounts of CF(3)H in common organic solvents such as tetrahydrofuran (THF), diethyl ether, and toluene. The methodology is widely applicable to a variety of silicon, boron, and sulfur-based electrophiles, as well as carbon-based electrophiles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%