Merck
  • Enzymic transfer of 6-modified D-galactosyl residues: synthesis of biantennary penta- and hepta-saccharides having two 6-deoxy-D-galactose residues at the nonreducing end and evaluation of 6-deoxy-D-galactosyl transfer to glycoprotein using bovine beta-(1-->4)-galactosyltransferase and UDP-6-deoxy-D-galactose.
Enzymic transfer of 6-modified D-galactosyl residues: synthesis of biantennary penta- and hepta-saccharides having two 6-deoxy-D-galactose residues at the nonreducing end and evaluation of 6-deoxy-D-galactosyl transfer to glycoprotein using bovine beta-(1-->4)-galactosyltransferase and UDP-6-deoxy-D-galactose.
Carbohydrate research (1995-04-19)
Y Kajihara, T Endo, H Ogasawara, H Kodama, H Hashimoto
ZUSAMMENFASSUNG
UDP-6-Deoxy-D-galactose and UDP-6-deoxy-6-fluoro-D-galactose were synthesized and their transfer to 2-acetamido-2-deoxy-D-glucose (N-acetyl-D-glucosamine) by beta-(1-->4)-galactosyltransferase was examined. The transfer rates of 6-deoxy-D-galactose and 6-deoxy-6-fluoro-D-galactose were 1.3 and 0.2% of that of D-galactosyl transfer, respectively. The 2-acetamido-4-O-(6-deoxy-beta-D-galactopyranosyl)-2-deoxy-D-glucopyranose (6'-deoxy-N-acetyllactosamine) and methyl 2-acetamido-4-O-(6-deoxy-6-fluoro-beta-D-galactopyranosyl)-2-deoxy-D- glucopyranoside (6'-deoxy-6'-fluoro-N-acetyllactosamine) were synthesized enzymatically in 30 and 59% yields, respectively. Further, 6-deoxy-D-galactose could be completely transferred to N-linked type biantennary oligosaccharides having two N-acetyl-D-glucosaminyl residues at the nonreducing end to give the corresponding penta- and hepta-saccharides in 55 and 57% yields, respectively. An assay of 6-deoxy-D-galactosyl transfer using asialo agalacto alpha 1-acid glycoprotein as an acceptor suggested that 6-deoxy-D-galactose was transferred to about 30% of the N-acetyl-D-glucosaminyl residues in the N-linked oligosaccharides of the glycoprotein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Galaktosyltransferase aus Kuhmilch, lyophilized powder