Merck
[Chemical constituents of Pilea cavaleriei subsp. cavaleriei].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-12-15)
Heng-chun Ren, Ri-dong Qin, Qing-ying Zhang, Wei Cheng, Hong Liang
ZUSAMMENFASSUNG
To investigate chemical constituents from folk herb Pilea cavaleriei subsp. cavaleriei. The compounds were separated and purified by silica gel, Sephadex LH-20 and the like. The structures were identified by spectral methods such as (1)H, (13)C-NMR and MS. Seventeen compounds were isolated and identified as benzoic acid (1), 4-hydroxy benzalde-hyde (2), coumaric acid(3), protocatechuic acid (4), gallic acid (5), 4-hydroxy benzoic acid (6), 3-indole carboxaldehyde (7), 3-indole carbo-xylicacid (8), 4-methyl-(1,2,3) -triazole(9), uracil(10), nicotinamide (11), (2S,E)-N-[2-hydroxy-2-(4-hydroxy phenyl) ethyl] ferulamide (12), (+) -dehydrovomifoliol (13), hentriantane (14), beta-sitosterol (15), palmitic acid (16), daucossterol (17) , respectively. All compounds were obtained from the genus for the first time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-50, Fine
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-25, Fine
Sigma-Aldrich
Sephadex® G-10, Medium
Sigma-Aldrich
Sephadex® G-50, Superfine
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-25, BioReagent, DNA grade, Molecular Biology, Superfine
Supelco
Sephadex® G-50, Medium
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-100, DNA grade, BioReagent, Molecular Biology, medium
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Sephadex® G-50, BioReagent, DNA grade, Molecular Biology, fine
Sigma-Aldrich
Sephadex® G-50, DNA grade, BioReagent, Molecular Biology, medium