Merck
  • Sildenafil, a phosphodiesterase type 5 inhibitor, reduces antidepressant-like activity of paroxetine in the forced swim test in mice.
Sildenafil, a phosphodiesterase type 5 inhibitor, reduces antidepressant-like activity of paroxetine in the forced swim test in mice.
Pharmacological reports : PR (2012-12-15)
Katarzyna Socała, Dorota Nieoczym, Elżbieta Wyska, Ewa Poleszak, Piotr Wlaź
ZUSAMMENFASSUNG
Sildenafil, a selective phosphodiesterase 5 (PDE5) inhibitor, has recently been reported to influence the antidepressant activity of some antidepressant drugs. The present study was undertaken to investigate the involvement of the nitric oxide/cyclic guanosine 3',5'-monophosphate/PDE5 (NO/cGMP/PDE5) signaling pathway in the antidepressant activity of paroxetine and to assess the interaction between paroxetine and sildenafil, in the forced swim test in mice. Swim trials were conducted by placing mice in glass cylinders filled with water for 6 min. Total behavioral immobility was measured during the last 4 min of the test. Changes in locomotor activity were measured with photoresistor actimeters. Serum and brain paroxetine concentrations were assayed by the HPLC method. Paroxetine at a dose of 1 mg/kg significantly decreased immobility time in the forced swim test, while sildenafil (5, 10 and 20 mg/kg) in a dose-dependent manner reduced the antidepressant activity of paroxetine. Pharmacokinetic studies did not show any significant changes in paroxetine concentration in serum and brain tissue as compared to paroxetine treatment alone. The results suggest that paroxetine may exert its antidepressant action by decreasing cGMP levels and sildenafil, as a drug which has the opposite effect on the processes mediated via the NO/cGMP/PDE5 signaling pathway, may decrease the efficacy of paroxetine. However, the co-administration of paroxetine with sildenafil resulted in a potent reduction (80%) of locomotor activity, which suggests that the reversal of antidepressant action of paroxetine may have been a result of locomotor deficits. Further studies are required to explain the mechanism underlying this phenomenon.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paroxetin -hydrochlorid Hemihydrat, ≥98% (HPLC), powder
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Paroxetin -hydrochlorid Hemihydrat, European Pharmacopoeia (EP) Reference Standard
Paroxetin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard