Merck
[Neuropharmacological potential of sigma1-receptors: new therapeutic possibilities].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2012-12-20)
O V Vorob'eva
ZUSAMMENFASSUNG
Sigma1-receptors are an independent class of intracellular receptors which are implicated in the regulation of cell bioenergetics that suggest their involvement in different neuropsychiatric diseases. Sigma1-receptors regulate different ion channels, including calcium channels, NMDA-receptors, and are involved in the neurotransmitter release, neurogenesis and synaptogenesis. These receptors recognize with a different affinity a variety of ligands of different structural classes with different therapeutic applications. High affinity to sigma1-receptors may play an important role in the mechanism of action of some antidepressants, in particular fluvoxamine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluvoxamin-Maleat, solid
Fluvoxamin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard