Merck
  • Endogenous repair by the activation of cell survival signalling cascades during the early stages of rat Parkinsonism.
Endogenous repair by the activation of cell survival signalling cascades during the early stages of rat Parkinsonism.
PloS one (2012-12-20)
Nga-Ping Lui, Liang-Wei Chen, Wing-Ho Yung, Ying-Shing Chan, Ken Kin-Lam Yung
ZUSAMMENFASSUNG
Here we report a previously unknown self repair mechanism during extremely early stages of rat Parkinsonism. Two important cell survival signaling cascades, Phosphatidylinositol-3 kinases (PI3K)/Akt pathway and extracellular signal-regulated kinase/mitogen-activated protein kinase (ERK/MAPK) pathway, could be responsible for this potential endogenous rescue system. In the 6-hydroxydopamine-lesioned rat, the phosphorylated p44/42 MAPK and its downstream target, the phosphorylated Bad at Ser 112, were up-regulated at post-lesion day 3 and lasted for a couple of weeks. Although the change in the phosphorylated Akt kinase was negligible throughout the studied period, its downstream target, the phosphorylated Bad at 136, was increased from post-lesion day 3 to post-lesion day 14. In the mean time, nestin-positive reactive astrocytes with low levels of brain-derived neurotrophic factor (BDNF) and glial cell line-derived neurotrophic factor (GDNF) appeared at post-lesion day 3 in 6-hydroxydopamine-lesioned rat. BDNF was expressed in both striatum and substantia nigra whereas GDNF was displayed in striatum only. At post-lesion day 14, nestin, BDNF and GDNF expressions were diminished. These neurotrophic factors were believed to initiate the above anti-apoptotic signal transduction cascades as we could see that their expression patterns were similar. The data strongly suggest that there is an endogenous repair effort by evoking the cell survival signaling and possibly via the releases of BDNF and GDNF from nestin-immunoreactive reactive astrocytes. ERK/MAPK pathway was proposed to be the key endogenous neuroprotective mechanisms, particularly in early stages of rat Parkinsonism. However, the self repair effort is only functional within an extremely short time window immediately after onset.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Tyrosin-Hydroxylase-Antikörper, Klon LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
6-Hydroxydopamin -hydrochlorid, ≥97% (titration), powder
Sigma-Aldrich
6-Hydroxydopamin -hydrobromid, 95%
Sigma-Aldrich
Anti-Nestin-Antikörper, Klon 401 (Ratte), clone rat-401, Chemicon®, from mouse