Merck
Distinct types of tumors exhibit differential grade of inflammation and angiogenesis in mice.
Microvascular research (2012-12-21)
C T R Viana, P P Campos, L A Carvalho, J M Cenedezi, L Lavall, M T P Lopes, M A N D Ferreira, S P Andrade
ZUSAMMENFASSUNG
Inflammation, angiogenesis and cytokine production are common features of almost, if not all tumors. However, the extent of these processes induced by different types of tumors has not been evaluated. We investigated the growth pattern of the experimental metastatic tumors, B16F10 melanoma, CT26.WT colon and 4T1 mammary cells inoculated in the flank of syngeneic mice and determined the degree of inflammation, angiogenesis, and production level of pro-inflammatory and pro-angiogenic cytokines within the tumors. In addition, we have analyzed vascular changes in the interface between the tumors and the adjacent cutaneous tissue and levels of relevant pro-inflammatory and pro-angiogenic cytokines systemically. The weight of tumors 15 days post-inoculation of 10(6) cells was markedly different. Melanomas were 2 and 10-fold heavier than colon and mammary tumors, respectively. Locally, CT26.WT tumor cells induced more vessels in cutaneous tissue adjacent to the tumors but systemically, the plasma levels of VEGF were higher (approximately 2-fold) in 4T1 tumor-bearing mice compared with the other two tumors. Mammary tumors presented the most prominent inflammatory content as assessed by a range of markers (inflammatory enzymes and cytokines). The vascular index, as determined by the intra-tumor amount of hemoglobin and number of vessels in hot spot areas, was also higher (approximately 2-fold) in melanomas compared with the other two tumors. These findings showing that distinct tumor types determine differential grade of inflammation, angiogenesis and host interaction in mice may provide new insights to tailor differential therapeutic approach based on the status of tumor biomarkers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%