Merck
MDMA can increase cortisol levels by 800% in dance clubbers.
Journal of psychopharmacology (Oxford, England) (2012-12-21)
Ac Parrott, J Lock, L Adnum, J Thome
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[Arg8]-Vasopressin -acetat (Salz), ≥95% (HPLC)
Sigma-Aldrich
(±)-3,4-Methylendioxymethamphetamin -hydrochlorid
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl