Merck
  • [Determination of fumonisins B1 and B2 in corn by high performance liquid chromatography with post-column derivatization method].
[Determination of fumonisins B1 and B2 in corn by high performance liquid chromatography with post-column derivatization method].
Se pu = Chinese journal of chromatography (2012-12-22)
Xiaoxu Zhang, Zhiyong Xiao, Hongyan Zhang, Lili Yang, Liyan Ma
ZUSAMMENFASSUNG
A high performance liquid chromatography-fluorescence detection with post-column derivatization method was developed to detect fumonisin B1 (FB1) and fumonisin B2 (FB2) in corn. Several factors, such as the pH of derivatization buffer, concentration and flow rate of derivatization reagents, excitation wavelength, emission wavelength, which affected the detection of fumonisins were optimized. The separation was performed on a ZORBAX SB C18 column operated at 40 degrees C with the gradient elution by two mobile phases of 0.1 mol/L sodium dihydrogen phosphate solution (pH 3.3) and methanol at a flow rate of 0.8 mL/min. The derivatization was performed at ambient temperature. The o-phthalaldehyde (OPA) flow rate was 0.4 mL/min. The results showed that the optimum conditions were pH 10.5 of the derivatization reagent, OPA concentration at 2 g/L, and excitation wavelength of 335 nm, emission wavelength of 440 nm. The linear plots of FB1 and FB2 were obtained between 0.2 to 20 mg/L, with the correlation coefficients above 0.999 for both FB1 and FB2. The limits of detection of fumonisins B1 and B2 were 0.02 mg/kg. The mean recoveries at the three spiked levels of 0.1 - 4.0 mg/kg were 82.5% - 89.8%. This method is accurate, simple, rapid and suitable for the determination of fumonisins B1 and B2 in corn.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fumonisin B1 aus Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor
Supelco
Fumonisin B1 -Lösung, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Fumonisin B2 -Lösung, ~50 μg/mL in acetonitrile: water (50:50), analytical standard