Merck
  • Effect of divalent metal ion impurities (Ba²⁺, Ca²⁺ and Mg²⁺) on the growth, structural and physical properties of KAP crystals.
Effect of divalent metal ion impurities (Ba²⁺, Ca²⁺ and Mg²⁺) on the growth, structural and physical properties of KAP crystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-25)
P Kanchana, A Elakkina Kumaran, Y Hayakawa, C Sekar
ZUSAMMENFASSUNG
Single crystals of potassium hydrogen phthalate (KAP), a semi-organic compound, have been grown by slow evaporation method at room temperature from aqueous solution in the presence of divalent metal ionic impurities Ba(2+), Ca(2+) and Mg(2+). Elemental analysis by inductively coupled plasma optical emission spectroscopy (ICP-OES) proves the incorporation of these impurities into the grown crystals. Powder X-ray diffraction studies confirmed the phase formation and metal ions doping into KAP crystals. Thermogravimetric-differential thermal analysis (TG-DTA) shows the onset decomposition temperatures to be 255, 238, 251 and 250°C for pure, Ba(2+), Ca(2+) and Mg(2+) doped KAP crystals respectively. Microhardness studies revealed that all the three doped crystals have improved hardness values than that of undoped KAP crystal. Ca-KAP crystal exhibited the highest second harmonic generation (SHG) conversion efficiency of 16 mV with the output power of nearly half of the standard potassium dihydrogen phosphate (KDP) crystal. The grown crystals were also subjected to Fourier transform infrared (FTIR) spectroscopy, ultra violet-visible-near infrared (UV-NIR) spectroscopy studies and dielectric studies. Among the three investigated metal ion impurities, Ca(2+) ion seem to have positive influence on the growth, mechanical, thermal, dielectric and SHG characteristics of KAP which makes it suitable for applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydrogenphthalat, BioXtra, ≥99.95%
Supelco
Kaliumphthalat, einbasisch, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Kaliumphthalat, einbasisch, ACS reagent, acidimetric standard
Sigma-Aldrich
Phthalsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumphthalat, einbasisch, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Kaliumphthalat, einbasisch, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Phthalsäure, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Supelco
Kaliumphthalat, einbasisch, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phthalsäure, reagent grade, 98%
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Phthalsäure Dikaliumsalz, 98%