Merck
  • Novel UV-spectrophotometric method for quantitative estimation of furazolidone using mixed hydrotropic agent.
Novel UV-spectrophotometric method for quantitative estimation of furazolidone using mixed hydrotropic agent.
Pakistan journal of pharmaceutical sciences (2012-12-25)
Nilesh Jain, Ruchi Jain, Deepak Kumar Jain, Rajesh Kumar Maheshwari, Surendra Jain
ZUSAMMENFASSUNG
A novel, eco friendly, accurate, sensitive, economic and safe spectrophotometric method was developed by application of mixed hydrotropy using 2 M sodium acetate, 8 M urea, 2 M niacinamide and 2 M sodium benzoate solution (25:25:25:25% V/V) as hydrotropic agent, for the solubalizing of poorly water-soluble Furazolidone (FZ) (solubility:- 3.64e-01 mg/mL in water). There were more than 32 times enhancements in the solubility of FZ were found in mixed hydrotropic solution as compared to solubilities in distilled water. FZ shows maximum absorbance at 360 nm where sodium acetate, urea, niacinamide, sodium benzoate and other tablets excipients did not show any absorbance above 300 nm, and thus no interference in the estimation was seen. FZ was obeyed Beers law in the concentration range of 10 to 50 μg/ml (r(2)=0.9992) in mixed hydrotropic solvent with mean recovery ranging from 97.32% to 98.9%. Proposed method is new, simple, economic, safe, rapid, accurate and reproducible and was validated according to ICH guidelines and values of accuracy, precision and other statistical analysis were found to be in good accordance with the prescribed values.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumacetat Trihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Natriumacetat Trihydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Supelco
Natriumbenzoat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumbenzoat, ReagentPlus®, 99%
Sigma-Aldrich
Natriumacetat Trihydrat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumbenzoat, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Natriumacetat Trihydrat, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumbenzoat, BioXtra, ≥99.5%
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumacetat Trihydrat, meets USP testing specifications
Sigma-Aldrich
Natriumacetat Trihydrat, BioXtra, ≥99.0%
Sigma-Aldrich
Furazolidon
Sigma-Aldrich
Natriumbenzoat, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%