Merck
  • Exploration of Vanilla pompona from the Peruvian Amazon as a potential source of vanilla essence: quantification of phenolics by HPLC-DAD.
Exploration of Vanilla pompona from the Peruvian Amazon as a potential source of vanilla essence: quantification of phenolics by HPLC-DAD.
Food chemistry (2012-12-26)
Helena Maruenda, Maria del Lujan Vico, J Ethan Householder, John P Janovec, Cristhian Cañari, Angelica Naka, Ana E Gonzalez
ZUSAMMENFASSUNG
This study provides the first chemical investigation of wild-harvested fruits of Vanilla pompona ssp. grandiflora (Lindl.) Soto-Arenas developed in their natural habitat in the Peruvian Amazon. Flowers were hand-pollinated and the resulting fruits were analysed at different developmental stages using an HPLC-DAD method validated for the quantification of glucovanillin and seven other compounds. The method showed satisfactory linearity (r(2)>0.9969), precision (coefficient of variation <2%), recoveries (70-100%), limit of detection (0.008-0.212 μg/ml), and limit of quantification (0.027-0.707 μg/ml). The evaluation of crude and enzyme-hydrolyzed Soxhlet-extracted samples confirmed the leading role of glucosides in fruit development. LC-ESI-MS studies corroborated the identities of four glucosides and seven aglycones, among them vanillin (5.7/100 g), 4-hydroxybenzyl alcohol (3.6/100 g), and anisyl alcohol (7.1/100 g) were found in high concentrations. The attractive flavor/aroma profile exhibited by wild V. pompona fruits supports studies focused on the development of this species as a specialty crop.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Vanillin Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Kalibriersubstanz von Mettler Toledo, ME 51143093, Vanillin, traceable to primary standards (LGC)
Vanillin, European Pharmacopoeia (EP) Reference Standard