Merck
  • In vitro anti-inflammatory effect of apigenin in the Helicobacter pylori-infected gastric adenocarcinoma cells.
In vitro anti-inflammatory effect of apigenin in the Helicobacter pylori-infected gastric adenocarcinoma cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-12-26)
Yuan-Chuen Wang, Kai-Ming Huang
ZUSAMMENFASSUNG
Infection with Helicobacter pylori causes extensive gastric epithelial cell inflammation which may progress to atrophic gastritis, intestinal metaplasia, and even gastric adenocarcinoma. Apigenin (4',5,7-trihydroxyflavone) is widely distributed in fruits and vegetables, and is a well-known antiinflammatory supplement with low cytotoxicity. In this study, we investigated the anti-inflammatory effects of apigenin in H. pylori-infected MKN45 cells, for which IκBα, cyclooxygenase-2 (COX-2), intercellular adhesion molecule-1 (ICAM-1), reactive oxygen species (ROS), interleukin-8 (IL-8), IL-6, IL-1β, and mucin-2 (MUC-2) expressions were examined. Apigenin treatments (9.3-74 μM) significantly increased the IκBα expression, and thus inhibited nuclear factor kappa B (NF-κB) activation, and the inflammatory factor (COX-2, ICAM-1, ROS, IL-6, and IL-8) expressions decreased. The ROS levels decreased partially based on the intrinsic scavenging property of apigenin. In summary, apigenin treatments effectively inhibited NF-κB activation and the related inflammatory factor expressions, as well as increased MUC-2 expression in the H. pylori-infected MKN45 cells. The compound shows great potential as a candidate agent for the inhibition of H. pylori-induced extensive gastric epithelial cell inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
Apigenin, analytical standard