Merck
  • p38MAPK and Rho-dependent kinase are involved in anoikis induced by anicequol or 25-hydroxycholesterol in DLD-1 colon cancer cells.
p38MAPK and Rho-dependent kinase are involved in anoikis induced by anicequol or 25-hydroxycholesterol in DLD-1 colon cancer cells.
Biochemical and biophysical research communications (2012-12-26)
Arowu R Tanaka, Kohji Noguchi, Hidesuke Fukazawa, Yasuhiro Igarashi, Hiroyuki Arai, Yoshimasa Uehara
ZUSAMMENFASSUNG
Anchorage-independent growth is evidence of the malignant transformation of cells. We previously reported the characterization of anicequol, a novel inhibitor of the anchorage-independent growth of tumor cells, and here we show that the effects of 25-hydroxycholesterol (25-HC) on colon cancer cells were very similar to those of anicequol. By analyzing the effects of inhibitors and performing RNA interference experiments, we found that p38 mitogen-activated protein kinase (p38MAPK) was involved in anicequol- and 25-HC-induced anoikis in DLD-1 cells. In addition, Rho-associated, coiled-coil containing protein kinase (ROCK) was also associated with anoikis induced by anicequol or 25-HC. Taken together, our findings suggest that activation of the p38MAPK and ROCK pathways might provide a new therapeutic strategy against cancer, and raise the possibility that tumor metastasis is influenced by 25-HC under physiological conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
25-Hydroxycholesterin, ≥98%