Merck
  • Signal transduction mechanism for potentiation by α1- and β2-adrenoceptor agonists of L-ascorbic acid-induced DNA synthesis and proliferation in primary cultures of adult rat hepatocytes.
Signal transduction mechanism for potentiation by α1- and β2-adrenoceptor agonists of L-ascorbic acid-induced DNA synthesis and proliferation in primary cultures of adult rat hepatocytes.
European journal of pharmacology (2012-12-29)
Hajime Moteki, Mitsutoshi Kimura, Katsuyoshi Sunaga, Tadashi Tsuda, Masahiko Ogihara
ZUSAMMENFASSUNG
We investigated the effects of α- and β-adrenoceptor agonists on L-ascorbic acid-induced hepatocyte DNA synthesis and proliferation in primary cultures of adult rat hepatocytes. The results showed that phenylephrine (10(-6) M) and metaproterenol (10(-6) M) alone did not induce hepatocyte DNA synthesis and proliferation. However, when combined with L-ascorbic acid (10(-6) M), these adrenoceptor agonists potentiated the hepatocyte DNA synthesis and proliferation induced by L-ascorbic acid. Then intracellular signal transduction mechanisms for the effects of phenylephrine and metaproterenol on L-ascorbic acid-induced hepatocyte mitogenesis were examined. Western blot analysis showed that phenylephrine and metaproterenol did not potentiate L-ascorbic acid-induced insulin-like growth factor I receptor tyrosine kinase phosphorylation. In contrast, they both significantly potentiated L-ascorbic acid-induced extracellular-signal regulated kinase-2 (ERK2) phosphorylation within 5 min. Moreover, cell-permeable second messenger analogs phorbol ester (10(-7) M) and 8-bromo cAMP (10(-7) M) mimicked the effects of phenylephrine and metaproterenol on L-ascorbic acid-induced ERK2 phosphorylation. The effects of these adrenoceptor agents were specifically antagonized by GF109203X and H-89, respectively. These results indicate that activation of ERK2 via protein kinas C and protein kinase A represents a mechanism for potentiation of L-ascorbic acid-induced hepatocyte DNA synthesis and proliferation in primary cultures of adult rat hepatocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zyklisches 8-Bromadenosin-3′,5′-Monophosphat Natriumsalz, ≥97% (HPLC), powder
Sigma-Aldrich
Zyklisches 8-Bromadenosin-3′,5′-monophosphat, ≥97% (HPLC)